1,1-Diphenylethylene 98%

Supplier: Thermo Scientific Chemicals

Synonyms: asym. Diphenylethene, Ethene-1,1-diyldibenzene, asym. Diphenylethylene

A14434-14 A14434-22 A14434-09 A14434-30
CAAAAA14434-14EA 169.06 CAD
CAAAAA14434-14 CAAAAA14434-22 CAAAA14434-09 CAAAA14434-30
1,1-Diphenylethylene 98%
1,1-Diphenylethylene

Purity: 98% CAS No.: 530-48-3 Molecular Formula: C14H12 Formula Weight: 180.25 Flash Point: 136°(276°F) Refractive Index: 1.6065 Melting Point: 5-7° Boiling Point: 270-271° Beilstein Reference: 1099062 Merck Reference: 14,3323 MDL No.: MFCD00008583

Formula: (C₆H₅)₂CCH₂
Boiling Pt: 270…271 °C
Melting Pt: 5…7 °C
Density: 1.023
Flash Pt: 136 °C(276 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008583
CAS Number: 530-48-3
EINECS: 208-482-6
Merck Index: 14,3323
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef9e2a018fc6e0a147414e06d2cdd5b51e949db2&vid=23a75fa0969db1a09c065b9fced36b49bc2d26df&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR