Indole 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14427-0B
A14427-18
A14427-30
CAAAA14427-0BEA
829.5
CAD
CAAAA14427-0B
CAAAA14427-18
CAAAA14427-30
Indole 99%
Indole
Purity: 99%
CAS No.: 120-72-9
Molecular Formula: C8H7N
Formula Weight: 117.15
Flash Point: 121°(249°F)
Odor: Unpleasant
pH: 6
Melting Point: 50-54°
Boiling Point: 253-254°
Beilstein Reference: 107693
Merck Reference: 14,4963
UN No.: 2811
MDL No.: MFCD00005607
Formula:
C₈H₇N Boiling Pt: 253…254 °C Melting Pt: 50…54 °C Density: 1.220 Flash Pt: 121 °C (249 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005607 CAS Number: 120-72-9 EINECS: 204-420-7 UN: 2811 ADR: 6.1,III Merck Index: 14,4963 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...