Methyl-3-hydroxybenzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl 3-Hydroxybenzoate, 3-Hydroxybenzoic acid methyl ester

A14425-14 A14425-22 A14425-36
CAAAA14425-14EA 91.01 CAD
CAAAA14425-14 CAAAA14425-22 CAAAA14425-36
Methyl-3-hydroxybenzoate 99%
Methyl 3-hydroxybenzoate

Purity: 99% CAS No.: 19438-10-9 Molecular Formula: C8H8O3 Formula Weight: 152.15 Melting Point: 70-72° Boiling Point: 280-281°/708mm MDL No.: MFCD00002295

Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 280…281 °C (708 mmHg)
Melting Pt: 70…72 °C
Storage Temperature: Ambient
MDL Number: MFCD00002295
CAS Number: 19438-10-9
EINECS: 243-071-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR