(±)-Linalool 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14424-0F
A14424-AE
A14424-AP
CAAAA14424-0FEA
682.56
CAD
CAAAA14424-0F
CAAAA14424-AE
CAAAA14424-AP
(±)-Linalool 97%
(±)-Linalool
Purity: 97%
CAS No.: 78-70-6
Molecular Formula: C10H18O
Formula Weight: 154.25
Flash Point: 76°(168°F)
Physical Form: Liquid
Density: 0.865
Refractive Index: 1.4620
Appearance: Colorless
Boiling Point: 199-200°
Beilstein Reference: 1721488
Merck Reference: 14,5495
MDL No.: MFCD00008906
Formula:
C₁₀H₁₈O Boiling Pt: 199…200 °C Density: 0.865 Flash Pt: 76 °C(168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008906 CAS Number: 78-70-6 EINECS: 201-134-4 Merck Index: 14,5495 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...