(4-Methoxyphenoxy)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(4-Methoxyphenoxy)acetic acid

A14419-09 A14419-18
CAAAA14419-09EA 72.21 CAD
CAAAA14419-09 CAAAA14419-18 CAAAA14419-30
(4-Methoxyphenoxy)acetic acid 98%
(4-Methoxyphenoxy)acetic acid

Purity: 98% CAS No.: 1877-75-4 Molecular Formula: C9H10O4 Formula Weight: 182.18 Melting Point: 109-113° Beilstein Reference: 1874579

Formula: (C₆H₄(OCH₃))OCH₂COOH
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 109…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00014360
CAS Number: 1877-75-4
EINECS: 217-513-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR