2,6-Dimethoxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14418-36
A14418-14
A14418-22
CAAAAA14418-36EA
614.62
CAD
CAAAAA14418-36
CAAAA14418-14
CAAAA14418-22
2,6-Dimethoxybenzoic acid 99%
2,6-Dimethoxybenzoic acid
Purity: 99%
CAS No.: 1466-76-8
Molecular Formula: C9H10O4
Formula Weight: 182.18
Odor: Odorless
Melting Point: ca 188° dec.
Beilstein Reference: 2212190
MDL No.: MFCD00002437
Formula:
C₉H₁₀O₄ MW: 182.18 g/mol Boiling Pt: 315 °C (1013 hPa) Melting Pt: ca 188 °C (dec.) Density: 1.365 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002437 CAS Number: 1466-76-8 EINECS: 215-985-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...