6-Nitroveratraldehyde 96%
Supplier: Thermo Scientific Chemicals
A14415-09
A14415-06
A14415-14
CAAAAA14415-09EA
115.97
CAD
CAAAAA14415-09
CAAAA14415-06
CAAAA14415-14
6-Nitroveratraldehyde 96%
6-Nitroveratraldehyde
Purity: 96%
CAS No.: 20357-25-9
Molecular Formula: C9H9NO5
Formula Weight: 211.17
Melting Point: 131-135°
Beilstein Reference: 395599
Formula:
C₉H₉NO₅ Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007134 CAS Number: 20357-25-9 EINECS: 243-762-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...