2-Nitro-4-(trifluoromethyl)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14413-09
A14413-18
A14413-30
CAAAA14413-09EA
66.52
CAD
CAAAA14413-09
CAAAA14413-18
CAAAA14413-30
2-Nitro-4-(trifluoromethyl)aniline 98%
2-Nitro-4-(trifluoromethyl)aniline
Purity: 98%
CAS No.: 400-98-6
Molecular Formula: C7H5F3N2O2
Formula Weight: 206.13
Melting Point: 105-109°
Beilstein Reference: 650808
UN No.: 2811
MDL No.: MFCD00007155
Formula:
C₇H₅F₃N₂O₂ Melting Pt: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007155 CAS Number: 400-98-6 EINECS: 206-926-3 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...