Salicyl alcohol 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Hydroxyphenyl)methanol, 2-(Hydroxymethyl)phenol, 2-Hydroxybenzyl alcohol, Saligenin

A14410-0B A14410-18 A14410-30
CAAAA14410-0BEA 1322.46 CAD
CAAAA14410-0B CAAAA14410-18 CAAAA14410-30
Salicyl alcohol 99%
Salicyl alcohol

Acts as a local anesthetic


Purity: 99% CAS No.: 90-01-7 Molecular Formula: C7H8O2 Formula Weight: 124.14 Melting Point: 83-87° Beilstein Reference: 1907195 Merck Reference: 14,8325 MDL No.: MFCD00004617

Formula: C₇H₈O₂
MW: 124.14 g/mol
Boiling Pt: 268
Melting Pt: 82
Storage Temperature: Ambient
MDL Number: MFCD00004617
CAS Number: 90-01-7
EINECS: 201-960-5
Merck Index: 14,8325

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR