Alizarine 94%

Supplier: Thermo Scientific Chemicals
Warning

A14404-0E A14404-22 A14404-36
CAAAA14404-0EEA 1655.84 CAD
CAAAA14404-0E CAAAA14404-22 CAAAA14404-36
Alizarine 94%
Alizarine

Purity: 94% CAS No.: 72-48-0 Molecular Formula: C14H8O4 Formula Weight: 240.21 Melting Point: 286-290° Boiling Point: ca 430° subl. Merck Reference: 14,251 MDL No.: MFCD00001201

Formula: C₁₄H₈O₄
Boiling Pt: ∼ 430 °C (subl.)
Melting Pt: 286…290 °C
Density: 1.54 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00001201
CAS Number: 72-48-0
EINECS: 200-782-5
Merck Index: 14,251

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR