Bromoacetic acid 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14403-0B A14403-0I A14403-30
CAAAA14403-0BEA 200.66 CAD
CAAAA14403-0B CAAAA14403-0I CAAAA14403-30
Bromoacetic acid 98+%
Bromoacetic acid

MDL: MFCD00002678 Beilstein: 506167 Soluble in water, alcohol


Purity: 98+% CAS No.: 79-08-3 Molecular Formula: C2H3BrO2 Formula Weight: 138.95 Flash Point: 110°(230°F) Odor: Pungent Boiling Point: 207-209° Solubility: Soluble in water, alcohol Beilstein Reference: 506167 Merck Reference: 14,1400 UN No.: 3425 MDL No.: MFCD00002678

Formula: BrCH₂COOH
Boiling Pt: 207…209 °C
Melting Pt: 44…49 °C
Density: 1.930
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002678
CAS Number: 79-08-3
EINECS: 201-175-8
UN: 3425
ADR: 8,II
Merck Index: 14,1400

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR