Phenyltrimethylammonium iodide 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N,N,N-Trimethylanilinium iodide, Trimethylphenylammonium iodide
A14402-22
A14402-36
CAAAA14402-22EA
118.82
CAD
CAAAA14402-22
CAAAA14402-36
Phenyltrimethylammonium iodide 99%
N,N,N-Trimethylanilinium iodide
Purity: 99%
CAS No.: 98-04-4
Molecular Formula: C6H5N(CH3)3I
Formula Weight: 263.12
Melting Point: ca 227° subl.
Beilstein Reference: 3916752
Merck Reference: 14,7318
UN No.: 2811
MDL No.: MFCD00011791
Formula:
C₉H₁₄IN MW: 263.12 g/mol Melting Pt: ∼ 227 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD00011791 CAS Number: 98-04-4 EINECS: 202-630-3 UN: 2811 Merck Index: 14,7318 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...