1,2-Cyclohexanedione 98+%
Supplier: Thermo Scientific Chemicals
A14401-06
A14401-14
CAAAA14401-06EA
153.1
CAD
CAAAA14401-06
CAAAA14401-14
1,2-Cyclohexanedione 98+%
1,2-Cyclohexanedione
Purity: 98+%
CAS No.: 765-87-7
Molecular Formula: C6H8O2
Formula Weight: 112.13
Storage Temperature: 2°C to 8°C
Flash Point: 84°(183°F)
Refractive Index: 1.5190
Melting Point: 35-38°
Boiling Point: 193-195°
Beilstein Reference: 507419
MDL No.: MFCD00001648
Formula:
C₆H₈O₂ MW: 112.13 g/mol Boiling Pt: 193…195 °C Melting Pt: 35…38 °C Density: 1.12 |
MDL Number:
MFCD00001648 CAS Number: 765-87-7 EINECS: 212-155-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...