(4-Methoxyphenyl)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Methoxyphenylacetic acid

A14399-06 A14399-22 A14399-36
CAAAAA14399-06EA 38.39 CAD
CAAAAA14399-06 CAAAA14399-22 CAAAA14399-36
(4-Methoxyphenyl)acetic acid 98+%
(4-Methoxyphenyl)acetic acid

Purity: 98+% CAS No.: 104-01-8 Molecular Formula: C9H10O3 Formula Weight: 166.18 Melting Point: 84-88° Boiling Point: 138-140°/3mm MDL No.: MFCD00004345

Formula: (C₆H₄(OCH₃))CH₂COOH
MW: 166.18 g/mol
Boiling Pt: 306
Melting Pt: 84…88 °C
Flash Pt: 193
Storage Temperature: Ambient
MDL Number: MFCD00004345
CAS Number: 104-01-8
EINECS: 203-166-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR