1-Hexadecanesulfonyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14396-09
A14396-18
CAAAA14396-09EA
146.94
CAD
CAAAA14396-09
CAAAA14396-18
CAAAA14396-30
1-Hexadecanesulfonyl chloride 98%
1-Hexadecanesulphonyl chloride
Purity: 98%
CAS No.: 38775-38-1
Molecular Formula: CH3(CH2)15SO2Cl
Formula Weight: 324.96
Melting Point: 56-58°
Beilstein Reference: 1789245
UN No.: 3261
MDL No.: MFCD00007459
Formula:
C₁₆H₃₃ClO₂S Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007459 CAS Number: 38775-38-1 EINECS: 254-123-1 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...