4-Hydroxycoumarin 98+%
Supplier: Thermo Scientific Chemicals
A14394-0E
A14394-22
A14394-36
CAAAAA14394-0EEA
1072.82
CAD
CAAAAA14394-0E
CAAAA14394-22
CAAAA14394-36
4-Hydroxycoumarin 98+%
4-Hydroxycoumarin
Purity: 98+%
CAS No.: 1076-38-6
Molecular Formula: C9H6O3
Formula Weight: 162.14
Odor: Odorless
Melting Point: ca 213° dec.
Beilstein Reference: 129768
MDL No.: MFCD00006856
Formula:
C₉H₆O₃ Boiling Pt: 349 °C (1013 hPa) Melting Pt: ∼ 213 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006856 CAS Number: 1076-38-6 EINECS: 214-060-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...