2,2-Dimethyl-1-propanol 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14390-14
A14390-22
A14390-36
CAAAA14390-14EA
78.21
CAD
CAAAA14390-14
CAAAA14390-22
CAAAA14390-36
2,2-Dimethyl-1-propanol 99%
2,2-Dimethyl-1-propanol
Purity: 99%
CAS No.: 75-84-3
Molecular Formula: (CH3)3CCH2OH
Formula Weight: 88.15
Flash Point: 36°(96°F)
Melting Point: 52-56°
Boiling Point: 111-114°
Beilstein Reference: 1730984
Merck Reference: 14,6457
UN No.: 1325
MDL No.: MFCD00004682
Formula:
C₅H₁₂O Boiling Pt: 111…114 °C Melting Pt: 52…56 °C Density: 0.818 Flash Pt: 36 °C (96 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004682 CAS Number: 75-84-3 EINECS: 200-907-3 UN: 1325 ADR: 4.1,III Merck Index: 14,6457 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...