1-Iodopropane 98+% stabilized
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Propyl iodide
Stabilization:
Stabilised with copper
A14388-22
A14388-36
CAAAA14388-22EA
73
CAD
CAAAA14388-22
CAAAA14388-36
1-Iodopropane 98+% stabilized
1-Iodopropane
Purity: 98+%
CAS No.: 107-08-4
Molecular Formula: C3H7I
Formula Weight: 169.99
Flash Point: 44°(111°F)
Refractive Index: 1.5050
Melting Point: -101°
Boiling Point: 101-102°
Beilstein Reference: 505937
Merck Reference: 14,7862
UN No.: 2392
MDL No.: MFCD00001095
Formula:
C₃H₇I MW: 169.99 g/mol Boiling Pt: 101…102 °C Melting Pt: –101 °C Density: 1.744 Flash Pt: 44 °C (111 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001095 CAS Number: 107-08-4 EINECS: 203-460-2 UN: 2392 ADR: 3,III Merck Index: 14,7862 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...