1,3-Difluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14387-09
A14387-18
A14387-30
CAAAA14387-09EA
100.96
CAD
CAAAA14387-09
CAAAA14387-18
CAAAA14387-30
1,3-Difluorobenzene 99%
1,3-Difluorobenzene
Purity: 99%
CAS No.: 372-18-9
Molecular Formula: C6H4F2
Formula Weight: 114.09
Flash Point: 2°(36°F)
Refractive Index: 1.4380
Odor: Characteristic
Melting Point: -59°
Boiling Point: 82-83°
Beilstein Reference: 1904537
UN No.: 1993
MDL No.: MFCD00000327
Formula:
C₆H₄F₂ Boiling Pt: 82…83 °C Melting Pt: –59 °C Density: 1.160 Flash Pt: 2 °C (36 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000327 CAS Number: 372-18-9 EINECS: 206-746-5 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...