Fluorobenzene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Phenylfluoride
A14384-0B
A14384-30
CAAAA14384-0BEA
320.74
CAD
CAAAA14384-0B
CAAAA14384-30
Fluorobenzene 99%
Fluorobenzene
Purity: 99%
CAS No.: 462-06-6
Molecular Formula: C6H5F
Formula Weight: 96.10
Flash Point: -15°(-5°F)
Refractive Index: 1.4660
Odor: Aromatic
Melting Point: -42°
Boiling Point: 84-85°
Beilstein Reference: 1236623
Merck Reference: 14,4170
UN No.: 2387
MDL No.: MFCD00000280
Formula:
C₆H₅F MW: 96.1 g/mol Boiling Pt: 84…85 °C Melting Pt: –42 °C Density: 1.025 Flash Pt: –15 °C (–5 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000280 CAS Number: 462-06-6 EINECS: 207-321-7 UN: 2387 ADR: 3,II Merck Index: 14,4170 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...