(Methoxymethyl)triphenylphosphonium chloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14380-14
A14380-22
A14380-36
CAAAA14380-14EA
97.01
CAD
CAAAA14380-14
CAAAA14380-22
CAAAA14380-36
(Methoxymethyl)triphenylphosphonium chloride 98+%
(Methoxymethyl)triphenylphosphonium chloride
Purity: 98+%
CAS No.: 4009-98-7
Molecular Formula: CH3OCH2P(C6H5)3Cl
Formula Weight: 342.81
Storage Temperature: 2°C to 8°C
Melting Point: ca 190° dec.
Beilstein Reference: 924215
MDL No.: MFCD00011800
Formula:
C₂₀H₂₀ClOP Melting Pt: ∼ 190 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00011800 CAS Number: 4009-98-7 EINECS: 223-664-5 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...