4-Nitrophenol 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1-Hydroxy-4-nitrobenzene, p-Nitrophenol
A14376-0B
A14376-0I
A14376-30
CAAAA14376-0BEA
115.34
CAD
CAAAA14376-0B
CAAAA14376-0I
CAAAA14376-30
4-Nitrophenol 99%
4-Nitrophenol
MDL: MFCD00007331
Beilstein: 1281877
Notes: 1.27.
Freely soluble in alcohol, chloroform, ether
Purity: 99%
CAS No.: 100-02-7
Molecular Formula: C6H5NO3
Formula Weight: 139.11
Flash Point: 169°(336°F)
Melting Point: 110-115°
Boiling Point: 279° dec.
Solubility: Freely soluble in alcohol, chloroform, ether
Beilstein Reference: 1281877
Merck Reference: 14,6621
UN No.: 1663
MDL No.: MFCD00007331
Formula:
C₆H₅NO₃ MW: 139.11 g/mol Boiling Pt: 279 (dec.) Melting Pt: 110…115 °C Density: 1.480 Flash Pt: 169 °C (336 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007331 CAS Number: 100-02-7 EINECS: 202-811-7 UN: 1663 ADR: 6.1 III Merck Index: 14,6621 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...