2-Fluorobenzonitrile 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: o-Fluorobenzonitrile

A14375-22 A14375-09 A14375-18 A14375-30
CAAAAA14375-22EA 382.36 CAD
CAAAAA14375-22 CAAAA14375-09 CAAAA14375-18 CAAAA14375-30
2-Fluorobenzonitrile 99%
2-Fluorobenzonitrile

Purity: 99% CAS No.: 394-47-8 Molecular Formula: C7H4FN Formula Weight: 121.11 Flash Point: 73°(163°F) Refractive Index: 1.5070 Odor: Almond-like Boiling Point: 196-198° Beilstein Reference: 2042184 MDL No.: MFCD00001773

Formula: C₇H₄FN
MW: 121.11 g/mol
Boiling Pt: 196…198 °C
Melting Pt: 35…37 °C
Density: 1.138
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001773
CAS Number: 394-47-8
EINECS: 206-897-7
UN: 3276
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR