3,5-Dimethoxybenzoic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14374-14
A14374-22
A14374-36
CAAAA14374-14EA
77.58
CAD
CAAAA14374-14
CAAAA14374-22
CAAAA14374-36
3,5-Dimethoxybenzoic acid 99%
3,5-Dimethoxybenzoic acid
Purity: 99%
CAS No.: 1132-21-4
Molecular Formula: C9H10O4
Formula Weight: 182.18
Odor: Odorless
Melting Point: 182-186°
Beilstein Reference: 511834
MDL No.: MFCD00002502
Formula:
C₉H₁₀O₄ Boiling Pt: 341 °C (1013 hPa) Melting Pt: 182…186 °C Density: 1,423 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002502 CAS Number: 1132-21-4 EINECS: 214-473-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...