4-tert-Butylbenzoyl chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-tert-Butylbenzoylchloride
A14373-14
A14373-22
CAAAA14373-14EA
103.96
CAD
CAAAA14373-14
CAAAA14373-22
4-tert-Butylbenzoyl chloride 98%
4-tert-Butylbenzoyl chloride
Purity: 98%
CAS No.: 1710-98-1
Molecular Formula: C11H13ClO
Formula Weight: 196.68
Flash Point: 87°(188°F)
Refractive Index: 1.5351
Boiling Point: 125-126°/10mm
Beilstein Reference: 775793
UN No.: 3265
MDL No.: MFCD00000695
Formula:
C₁₁H₁₃ClO MW: 196.68 g/mol Boiling Pt: 125…126 °C/10 mm Density: 1.080 Flash Pt: 87 °C (188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000695 CAS Number: 1710-98-1 EINECS: 216-973-1 UN: 2922 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...