4,4'-Difluorobenzhydrol 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: 4,4'-Difluorophenyl phenyl carbinol, 4,4'-Difluorobenzhydryl alcohol, 4,4'-Difluoro-α-phenylbenzyl alcohol, Bis(4-fluorophenyl)carbinol, 4,4'-Difluorodiphenylmethanol, Bis(4-fluorophenyl)methanol

A14370-06 A14370-14 A14370-22
CAAAA14370-06EA 33.65 CAD
CAAAA14370-06 CAAAA14370-14 CAAAA14370-22
4,4'-Difluorobenzhydrol 98+%
4,4'-Difluorobenzhydrol

Purity: 98+% CAS No.: 365-24-2 Molecular Formula: C13H10F2O Formula Weight: 220.22 Flash Point: 110°(230°F) Melting Point: 45-49° Boiling Point: 142-144°/3mm Beilstein Reference: 2332258 MDL No.: MFCD00000357

Formula: C₁₃H₁₀F₂O
Boiling Pt: 142…144 °C/3 mm
Melting Pt: 45…49 °C
Density: 1 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000357
CAS Number: 365-24-2
EINECS: 206-671-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR