Trifluoroacetic acid ≥99.5% for biochemistry

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: TFA

A14365-0B A14365-18 A14365-30
CAAAA14365-0BEA 808.96 CAD
CAAAA14365-0B CAAAA14365-18 CAAAA14365-30
Trifluoroacetic acid ≥99.5% for biochemistry
Trifluoroacetic acid

Used to cleave peptide blocking groups


Purity: 99.5+% CAS No.: 76-05-1 Molecular Formula: CF3CO2H Formula Weight: 114.02 Refractive Index: 1.2840 Odor: Pungent Boiling Point: 72-73° Beilstein Reference: 742035 Merck Reference: 14,9681 UN No.: 2699 MDL No.: MFCD00004169

Formula: C₂HF₃O₂
MW: 114.02 g/mol
Boiling Pt: 71.8
Melting Pt: -15
Density: 1.480
Storage Temperature: Ambient
MDL Number: MFCD00004169
CAS Number: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8 I
Merck Index: 14,9681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR