2',3',4'-Trihydroxyacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A14364-06 A14364-14 A14364-22
CAAAA14364-06EA 74.73 CAD
CAAAA14364-06 CAAAA14364-14 CAAAA14364-22
2',3',4'-Trihydroxyacetophenone 98%
2',3',4'-Trihydroxyacetophenone

Purity: 98% CAS No.: 528-21-2 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 169-172° Merck Reference: 14,4342 MDL No.: MFCD00002193

Formula: C₈H₈O₄
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00002193
CAS Number: 528-21-2
EINECS: 208-430-2
Merck Index: 14,4342

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR