Acetanilide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14361-22
A14361-0E
A14361-36
CAAAAA14361-22EA
58.46
CAD
CAAAAA14361-22
CAAAA14361-0E
CAAAA14361-36
Acetanilide 98%
Acetanilide
Purity: 98%
CAS No.: 103-84-4
Molecular Formula: C8H9NO
Formula Weight: 135.17
Flash Point: 161°(321°F)
Physical Form: Crystals
Density: 1.121
Melting Point: 113-116°
Boiling Point: 304-305°
Beilstein Reference: 606468
Merck Reference: 14,50
MDL No.: MFCD00008674
Formula:
C₈H₉NO Boiling Pt: 304…305 °C Melting Pt: 113…116 °C Density: 1.121 Flash Pt: 161 °C(321 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008674 CAS Number: 103-84-4 EINECS: 203-150-7 Merck Index: 14,50 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...