2-Bromo-p-xylene 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14358-14
A14358-22
CAAAA14358-14EA
81.69
CAD
CAAAA14358-14
CAAAA14358-22
CAAAA14358-36
2-Bromo-p-xylene 98+%
2-Bromo-p-xylene
Purity: 98+%
CAS No.: 553-94-6
Molecular Formula: C8H9Br
Formula Weight: 185.07
Flash Point: 79°(174°F)
Refractive Index: 1.5510
Melting Point: 9-10°
Boiling Point: 199-201°
Beilstein Reference: 2040350
MDL No.: MFCD00000074
Formula:
C₈H₉Br Boiling Pt: 199…201 °C Melting Pt: 9…10 °C Density: 1.347 Flash Pt: 79 °C(174 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000074 CAS Number: 553-94-6 EINECS: 209-055-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...