1-Adamantanecarbonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

A14352-03 A14352-06 A14352-14
CAAAA14352-03EA 62.57 CAD
CAAAA14352-03 CAAAA14352-06 CAAAA14352-14
1-Adamantanecarbonitrile 97%
1-Adamantanecarbonitrile

Purity: 97% CAS No.: 23074-42-2 Molecular Formula: C11H15N Formula Weight: 161.25 Melting Point: 193-196° UN No.: 3439 MDL No.: MFCD00074731

Formula: C₁₁H₁₅N
Boiling Pt: 275 °C (1013 hPa)
Melting Pt: 193…196 °C
Density: 1.13 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00074731
CAS Number: 23074-42-2
EINECS: 245-413-9
UN: 3439
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR