Thiophene-2-carbaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14344-22
A14344-36
A14344-0B
CAAAAA14344-22EA
84.06
CAD
CAAAAA14344-22
CAAAAA14344-36
CAAAA14344-0B
Thiophene-2-carbaldehyde 98+%
Thiophene-2-carbaldehyde
Purity: 98+%
CAS No.: 98-03-3
Molecular Formula: C5H4OS
Formula Weight: 112.15
Storage Temperature: 2°C to 8°C
Flash Point: 78°(172°F)
Refractive Index: 1.5900
Boiling Point: 196-198°
Beilstein Reference: 105819
MDL No.: MFCD00005429
Formula:
C₅H₄OS Boiling Pt: 196…198 °C Melting Pt: ? 10 °C Density: 1,220 Flash Pt: 78 °C(172 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005429 CAS Number: 98-03-3 EINECS: 202-629-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...