N-phenylanthranilic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(Phenylamino)benzoic acid, Fenamic acid

A14340-14 A14340-22 A14340-36
CAAAA14340-14EA 66.2 CAD
CAAAA14340-14 CAAAA14340-22 CAAAA14340-36
N-phenylanthranilic acid 99%
N-phenylanthranilic acid

Purity: 99% CAS No.: 91-40-7 Molecular Formula: C13H11NO2 Formula Weight: 213.24 Melting Point: 185-188° Beilstein Reference: 1456607 Merck Reference: 14,7273 MDL No.: MFCD00002421

Formula: C₁₃H₁₁NO₂
MW: 213.24 g/mol
Boiling Pt: 375
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00002421
CAS Number: 91-40-7
EINECS: 202-066-8
Merck Index: 14,7273

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR