2,3-Butanedione monoxime 99%
Supplier: Thermo Scientific Chemicals
A14339-0E
A14339-22
A14339-36
CAAAA14339-0EEA
1643.2
CAD
CAAAA14339-0E
CAAAA14339-22
CAAAA14339-36
2,3-Butanedione monoxime 99%
2,3-Butanedione monoxime
For determination of urea
Purity: 99%
CAS No.: 57-71-6
Molecular Formula: C4H7NO2
Formula Weight: 101.11
Storage Temperature: 2°C to 8°C
Melting Point: 74-77°
Boiling Point: 185-186°
Beilstein Reference: 605582
MDL No.: MFCD00002116
Formula:
C₄H₇NO₂ Boiling Pt: 185…186 °C Melting Pt: 74…77 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002116 CAS Number: 57-71-6 EINECS: 200-348-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...