5-Chloroanthranilic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Amino-5-chlorobenzoic acid

A14329-06 A14329-14 A14329-22
CAAAA14329-06EA 78.37 CAD
CAAAA14329-06 CAAAA14329-14 CAAAA14329-22
5-Chloroanthranilic acid 98%
5-Chloroanthranilic acid

Purity: 98% CAS No.: 635-21-2 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 208° dec. Beilstein Reference: 639027

Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: ca 208 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007838
CAS Number: 635-21-2
EINECS: 211-230-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR