4-Benzylpyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14326-22
A14326-36
A14326-18
CAAAAA14326-22EA
89.59
CAD
CAAAAA14326-22
CAAAAA14326-36
CAAAA14326-18
4-Benzylpyridine 99%
4-Benzylpyridine
Purity: 99%
CAS No.: 2116-65-6
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 115°(239°F)
Refractive Index: 1.5805
Melting Point: 9-11°
Boiling Point: 286-287°
Beilstein Reference: 115831
MDL No.: MFCD00006443
Formula:
C₁₂H₁₁N MW: 169.23 g/mol Boiling Pt: 286…287 °C Melting Pt: 9…11 °C Density: 1.061 Flash Pt: 115 °C(239 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006443 CAS Number: 2116-65-6 EINECS: 218-319-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...