3-(3,4-Dimethoxyphenyl)propionic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid

A14325-06 A14325-14
CAAAAA14325-22EA 652.45 CAD
CAAAAA14325-22 CAAAA14325-06 CAAAA14325-14
3-(3,4-Dimethoxyphenyl)propionic acid 98%
3-(3,4-Dimethoxyphenyl)propionic acid

Purity: 98% CAS No.: 2107-70-2 Molecular Formula: C11H14O4 Formula Weight: 210.23 Melting Point: 96-99° Beilstein Reference: 2696272

Formula: C₁₁H₁₄O₄
MW: 210.23 g/mol
Boiling Pt: 344
Melting Pt: 102
Storage Temperature: Ambient
MDL Number: MFCD00002774
CAS Number: 2107-70-2
EINECS: 218-288-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR