1,9-Dibromononane 97%

Supplier: Thermo Scientific Chemicals
Warning

A14324-14 A14324-09 A14324-18
CAAAAA14324-14EA 147.41 CAD
CAAAAA14324-14 CAAAA14324-09 CAAAA14324-18
1,9-Dibromononane 97%
1,9-Dibromononane

Purity: 97% CAS No.: 4549-33-1 Molecular Formula: C9H18Br2 Formula Weight: 286.06 Flash Point: 110°(230°F) Refractive Index: 1.4965 Boiling Point: 285-288° Beilstein Reference: 1737598 MDL No.: MFCD00000279

Formula: BrH₂C(CH₂)₇CH₂Br
Boiling Pt: 285…288 °C
Melting Pt: –10 °C
Density: 1.410
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000279
CAS Number: 4549-33-1
EINECS: 224-913-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR