N-Iodosuccinimide 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14320-06
A14320-14
A14320-22
CAAAA14320-06EA
83.9
CAD
CAAAA14320-06
CAAAA14320-14
CAAAA14320-22
N-Iodosuccinimide 97%
N-Iodosuccinimide
Purity: 97%
CAS No.: 516-12-1
Molecular Formula: C4H4INO2
Formula Weight: 224.99
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Density: 2.245
Sensitivity: Moisture and Light Sensitive
Appearance: White to pale brown
Melting Point: ca 196° dec.
Beilstein Reference: 113917
Merck Reference: 14,5045
MDL No.: MFCD00005512
Formula:
C₄H₄INO₂ Boiling Pt: 356 °C (1013 hPa) Melting Pt: ∼ 196 °C (dec.) Density: 2.245 Storage Temperature: Refrigerator |
MDL Number:
MFCD00005512 CAS Number: 516-12-1 EINECS: 208-221-6 Merck Index: 14,5045 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...