4-Bromo-2-methyl-6-nitroaniline 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-6-nitro-o-toluidine, 2-Amino-5-bromo-3-nitrotoluene
A14319-06
A14319-14
CAAAA14319-03EA
39.52
CAD
CAAAA14319-03
CAAAA14319-06
CAAAA14319-14
4-Bromo-2-methyl-6-nitroaniline 97%
4-Bromo-2-methyl-6-nitroaniline
Purity: 97%
CAS No.: 77811-44-0
Molecular Formula: C7H7BrN2O2
Formula Weight: 231.06
Melting Point: 143-147°
Beilstein Reference: 3530524
Formula:
C₇H₇BrN₂O₂ Melting Pt: 143…147 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052919 CAS Number: 77811-44-0 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...