(3-Methoxyphenyl)acetonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

A14317-06 A14317-14 A14317-22
CAAAA14317-06EA 67.47 CAD
CAAAA14317-06 CAAAA14317-14 CAAAA14317-22
(3-Methoxyphenyl)acetonitrile 99%
(3-Methoxyphenyl)acetonitrile

Purity: 99% CAS No.: 19924-43-7 Molecular Formula: C9H9NO Formula Weight: 147.18 Flash Point: 98°(208°F) Refractive Index: 1.5312 Boiling Point: 278-282° Beilstein Reference: 1865539 UN No.: 3276 MDL No.: MFCD00001910

Formula: C₉H₉NO
Boiling Pt: 278…282 °C
Density: 1.082
Flash Pt: 98 °C (208 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001910
CAS Number: 19924-43-7
EINECS: 243-428-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR