3-(2-Methoxyphenyl)propionic acid 98+%

Supplier: Thermo Scientific Chemicals

A14314-06 A14314-14 A14314-22
CAAAA14314-06EA 29.23 CAD
CAAAA14314-06 CAAAA14314-14 CAAAA14314-22
3-(2-Methoxyphenyl)propionic acid 98+%
3-(2-Methoxyphenyl)propionic acid

Purity: 98+% CAS No.: 6342-77-4 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 85-88° Beilstein Reference: 2097012

Formula: C₁₀H₁₂O₃
Boiling Pt: 302 °C (1013 hPa)
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD00002772
CAS Number: 6342-77-4
EINECS: 228-738-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR