α-Bromo-3-nitrotoluene 98+%

Supplier: Thermo Scientific Chemicals
Danger

A14310-06 A14310-14 A14310-22
CAAAA14310-06EA 60.67 CAD
CAAAA14310-06 CAAAA14310-14 CAAAA14310-22
α-Bromo-3-nitrotoluene 98+%
α-Bromo-3-nitrotoluene

Purity: 98+% CAS No.: 3958-57-4 Molecular Formula: C7H6BrNO2 Formula Weight: 216.04 Melting Point: 56-59° Boiling Point: 153-154°/7mm UN No.: 3261 MDL No.: MFCD00007271

Formula: C₇H₆BrNO₂
Boiling Pt: 153…154 °C/7 mm
Melting Pt: 56…59 °C
Storage Temperature: Ambient
MDL Number: MFCD00007271
CAS Number: 3958-57-4
EINECS: 223-557-3
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR