1,1,3,3-Tetramethyldisilazane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14304-09
A14304-18
CAAAA14304-09EA
120.87
CAD
CAAAA14304-09
CAAAA14304-18
1,1,3,3-Tetramethyldisilazane 97%
1,1,3,3-Tetramethyldisilazane
Purity: 97%
CAS No.: 15933-59-2
Molecular Formula: C4H15NSi2
Formula Weight: 133.34
Flash Point: -3°(26°F)
Refractive Index: 1.4055
Odor: Ammonia-like
Boiling Point: 99-100°
Beilstein Reference: 741869
UN No.: 2924
MDL No.: MFCD00025626
Formula:
C₄H₁₅NSi₂ Boiling Pt: 99…100 °C Density: 0.768 Flash Pt: –3 °C (26 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025626 CAS Number: 15933-59-2 EINECS: 240-072-2 UN: 2924 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...