4-Bromo-m-toluic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-3-methylbenzoic acid, 2-Bromo-5-carboxytoluene, 4-Bromo-m-toluic acid
A14302-06
A14302-14
CAAAA14302-03EA
78.98
CAD
CAAAA14302-03
CAAAA14302-06
CAAAA14302-14
4-Bromo-m-toluic acid 98%
4-Bromo-m-toluic acid
Purity: 98%
CAS No.: 7697-28-1
Molecular Formula: C8H7BrO2
Formula Weight: 215.05
Melting Point: 215-218°
Beilstein Reference: 2356649
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol Melting Pt: 215…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00039526 CAS Number: 7697-28-1 EINECS: 231-713-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...