5-Nitrovanillin 97%
Supplier: Thermo Scientific Chemicals
A14296-22
A14296-06
A14296-14
CAAAAA14296-22EA
470.84
CAD
CAAAAA14296-22
CAAAA14296-06
CAAAA14296-14
5-Nitrovanillin 97%
5-Nitrovanillin
Purity: 97%
CAS No.: 6635-20-7
Molecular Formula: C8H7NO5
Formula Weight: 197.15
Melting Point: 174-178°
Beilstein Reference: 1973746
Formula:
C₈H₇NO₅ Melting Pt: 174…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007118 CAS Number: 6635-20-7 EINECS: 229-633-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...