Benzyltriethylammonium bromide 98+%

Supplier: Thermo Scientific Chemicals
Warning

A14291-14 A14291-22 A14291-36
CAAAA14291-14EA 46.61 CAD
CAAAA14291-14 CAAAA14291-22 CAAAA14291-36
Benzyltriethylammonium bromide 98+%
Benzyltriethylammonium bromide

Purity: 98+% CAS No.: 5197-95-5 Molecular Formula: (CH3CH2)3N(CH2C6H5)Br Formula Weight: 272.24 Melting Point: ca 200° dec. Beilstein Reference: 3739113

Formula: C₁₃H₂₂BrN
MW: 272.23 g/mol
Melting Pt: ∼ 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011822
CAS Number: 5197-95-5
EINECS: 225-986-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR