Methyl-4-hydroxybenzoate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14289-0B
A14289-0I
A14289-30
CAAAA14289-0BEA
192.76
CAD
CAAAA14289-0B
CAAAA14289-0I
CAAAA14289-30
Methyl-4-hydroxybenzoate 99%
Methyl 4-hydroxybenzoate
Purity: 99%
CAS No.: 99-76-3
Molecular Formula: C8H8O3
Formula Weight: 152.15
Odor: Odorless
pH: 5.8
Melting Point: 125-128°
Boiling Point: ca 280° dec.
Beilstein Reference: 509801
Merck Reference: 14,6107
MDL No.: MFCD00002352
Formula:
HOC₆H₄CO₂CH₃ Boiling Pt: ∼ 280 °C (dec.) Melting Pt: 125…128 °C Density: 1.1208 g/cm³ (137 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002352 CAS Number: 99-76-3 EINECS: 202-785-7 Merck Index: 14,6107 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...