1,1-Cyclobutanedicarboxylic acid 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A14288-06
A14288-14
A14288-22
CAAAA14288-06EA
76.47
CAD
CAAAA14288-06
CAAAA14288-14
CAAAA14288-22
1,1-Cyclobutanedicarboxylic acid 99%
1,1-Cyclobutanedicarboxylic acid
Purity: 99%
CAS No.: 5445-51-2
Molecular Formula: C6H8O4
Formula Weight: 144.13
Melting Point: ca 158° dec.
Beilstein Reference: 2046031
UN No.: 3261
MDL No.: MFCD00001325
Formula:
C₆H₈O₄ Boiling Pt: 663 °C (1013 hPa) Melting Pt: ∼ 158 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001325 CAS Number: 5445-51-2 EINECS: 226-651-2 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...