2-Amino-5-bromopyridine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A14280-06
A14280-14
A14280-22
CAAAA14280-06EA
53.25
CAD
CAAAA14280-06
CAAAA14280-14
CAAAA14280-22
2-Amino-5-bromopyridine 98%
2-Amino-5-bromopyridine
Purity: 98%
CAS No.: 1072-97-5
Molecular Formula: C5H5BrN2
Formula Weight: 173.01
Melting Point: 135-140°
Beilstein Reference: 108737
Formula:
C₅H₅BrN₂ Boiling Pt: 244 °C (1013 hPa) Melting Pt: 135…140 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006323 CAS Number: 1072-97-5 EINECS: 214-019-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...